Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-DPCA ethyl ester | 86443-19-8 | 99.48% | 268.27 | 1 ML
SDP

Catalog No. 5000351117
Encompass_Preferred

1,4-DPCA ethyl ester is the ethyl ester of 1,4-DPCA and functions as an inhibitor of factor inhibiting HIF (FIH). This tricyclic compound is intended for research use only.

  • Inhibitor of factor inhibiting HIF (FIH)
  • Tricyclic compound
  • For research use only
  • Purity: 99.48%
  • Molecular weight: 268.27
  • Appearance: Off-white to light yellow solid
  • Chemical formula: C15H12N2O3

Catalog No. 50-003-51117 Supplier Medchemexpress LLC Supplier No. HY13723910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-DPCA ethyl ester is the ethyl ester of 1,4-DPCA and functions as an inhibitor of factor inhibiting HIF (FIH). This tricyclic compound is intended for research use only.

  • Inhibitor of factor inhibiting HIF (FIH)
  • Tricyclic compound
  • For research use only
  • Purity: 99.48%
  • Molecular weight: 268.27
  • Appearance: Off-white to light yellow solid
  • Chemical formula: C15H12N2O3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.