Learn More
Medchemexpress LLC 1,4-DPCA ethyl ester | 86443-19-8 | 99.5% | 268.27 | 25 MG
SDP
Catalog No. 5000348906
Encompass_Preferred
1,4-DPCA ethyl ester is the ethyl ester of 1,4-DPCA, a tricyclic compound that inhibits factor inhibiting HIF (FIH). This product appears as an off-white to light yellow solid.
  • Inhibits factor inhibiting HIF (FIH)
  • Solid form
  • Off-white to light yellow color
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-48906 Supplier Medchemexpress LLC Supplier No. 25MGHY137239
1,4-DPCA ethyl ester is the ethyl ester of 1,4-DPCA, a tricyclic compound that inhibits factor inhibiting HIF (FIH). This product appears as an off-white to light yellow solid.
  • Inhibits factor inhibiting HIF (FIH)
  • Solid form
  • Off-white to light yellow color
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month