Learn More
Medchemexpress LLC 1,4-DPCA ethyl ester | 86443-19-8 | 99.5% | 268.27 | 50 MG
SDP
Catalog No. 5000351728
Encompass_Preferred
1,4-DPCA ethyl ester is the ethyl ester of 1,4-DPCA, a tricyclic compound that can inhibit factor inhibiting HIF (FIH). It is for research use only.
  • Ethyl ester of 1,4-DPCA
  • Inhibits factor inhibiting HIF (FIH)
  • Tricyclic compound
Catalog No. 50-003-51728 Supplier Medchemexpress LLC Supplier No. HY13723950MG
1,4-DPCA ethyl ester is the ethyl ester of 1,4-DPCA, a tricyclic compound that can inhibit factor inhibiting HIF (FIH). It is for research use only.
  • Ethyl ester of 1,4-DPCA
  • Inhibits factor inhibiting HIF (FIH)
  • Tricyclic compound