Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Ethoxyphenyl)-5-mercapto-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. E06685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E06685G Supplier TCI America Supplier No. E06685G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15182-68-0
Molecular Formula C9H10N4OS
Molecular Weight (g/mol) 222.27
MDL Number MFCD02252889
InChI Key ASFXKDBHBVHSLT-UHFFFAOYSA-N
Synonym 1-(4-Ethoxyphenyl)-1H-tetrazole-5-thiol
PubChem CID 853337
IUPAC Name 1-(4-ethoxyphenyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
SMILES CCOC1=CC=C(C=C1)N1NN=NC1=S

Specifications

Melting Point 160°C
Color White
Quantity 5 g
UN Number 1325
Formula Weight 222.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Ethoxyphenyl)-5-mercapto-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.