Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(4-fluorophenyl)-2-methylpropan-2-amine | 1200-27-7 | MFCD00082702 | 97.0% | 167.22 g/mol | C10H14FN | 1 G
SDP

Catalog No. 5000282089
Encompass_Preferred

1-(4-Fluorophenyl)-2-methylpropan-2-amine is an organic research chemical (CAS 1200-27-7) used as a building block in organic synthesis and chemical research. It has formula C10H14FN and a molecular weight of 167.22 g/mol. The compound is sold for laboratory research use only; purity is lot-dependent and should be confirmed from the supplier certificate of analysis.

  • Used as a synthetic building block in organic chemistry.
  • Chemical formula C10H14FN.
  • Molecular weight 167.22 g/mol.
  • Supplied in small pack sizes suitable for research.
  • Not for human or clinical use; for research use only.
  • Verify lot-specific purity via certificate of analysis.

Catalog No. 50-002-82089 Supplier Medchemexpress LLC Supplier No. HYW3307021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(4-Fluorophenyl)-2-methylpropan-2-amine is an organic research chemical (CAS 1200-27-7) used as a building block in organic synthesis and chemical research. It has formula C10H14FN and a molecular weight of 167.22 g/mol. The compound is sold for laboratory research use only; purity is lot-dependent and should be confirmed from the supplier certificate of analysis.

  • Used as a synthetic building block in organic chemistry.
  • Chemical formula C10H14FN.
  • Molecular weight 167.22 g/mol.
  • Supplied in small pack sizes suitable for research.
  • Not for human or clinical use; for research use only.
  • Verify lot-specific purity via certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.