Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(4-fluorophenyl)-2-methylpropan-2-amine | 1200-27-7 | MFCD00082702 | >=98.0% | 167.22 g/mol | C10H14FN | 250 MG
SDP

Supplier:  Medchemexpress LLC HYW330702250MG

Encompass_Preferred

1-(4-fluorophenyl)-2-methylpropan-2-amine is a small organic tertiary amine used as a synthetic building block and reagent in chemical research. It is intended for research use only and is supplied in multiple pack sizes for laboratory applications.

  • Research use only (not for human use).
  • CAS number 1200-27-7.
  • Molecular formula C10H14FN; molecular weight 167.22 g/mol.
  • Available in multiple pack sizes including 250 mg.
  • High purity: >=98.0%.
  • Identifiers include SMILES and MFCD00082702.

Catalog No. 50-002-82522


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.