Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)cyclopentanecarbonitrile, 99%, Thermo Scientific™

Catalog No. AC304830050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304830050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC304830050 Supplier Thermo Scientific Chemicals Supplier No. 304830050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83706-50-7
Molecular Formula C12H12FN
Molecular Weight (g/mol) 189.09
MDL Number MFCD00800616
InChI Key MAGOCEYAQAZRNE-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl cyclopentanecarbonitrile, 1-4-fluorophenyl cyclopentane-1-carbonitrile, 1-4-fluoro-phenyl-cyclopentanecarbonitrile, cyclopentanecarbonitrile,1-4-fluorophenyl
PubChem CID 2733239
IUPAC Name 1-(4-fluorophenyl)cyclopentane-1-carbonitrile
SMILES C1CCC(C1)(C#N)C2=CC=C(C=C2)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5140 to 1.516
Quantity 5 g
Formula Weight 189.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-(4-Fluorophenyl)cyclopentanecarbonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 280-526-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.