Learn More
Medchemexpress LLC 1-(4-fluorophenyl)-N-[1-(2-methylsulfonylpyridin-4-yl)cyclopropyl]pyrazolo[3,4-c]pyridine-4-carboxam | 1295298-26-8 | 98.9% | 50 MG

Supplier: Medchemexpress LLC HY12058850MG
BI 639667 is a small-molecule chemical probe and potent antagonist of the chemokine receptor CCR1. It has been characterized in multiple in vitro assays and reported to have low-nanomolar activity; key identifiers include CAS 1295298-26-8 and molecular formula C22H18FN5O3S.
- Potent CCR1 antagonist with low-nanomolar activity in cell-based and binding assays.
- Used as a chemical probe for CCR1 target validation and pharmacology studies.
- Chemical formula C22H18FN5O3S and molecular weight ~451.47 Da.
- High reported purity (98.87%) for supplied solid material.
- Available in multiple sizes and as a 10 mM solution in DMSO for screening.
- Recommended storage: powder and in-solvent conditions per supplier guidance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.