Learn More
Medchemexpress LLC 1-(4-fluorophenyl)-N-((2-(methylsulfonyl)pyridin-4-yl)methyl)-1H-pyrazolo[3,4-c]pyridine | 1220026-26-5 | 99.7% | 425.44 | C20H16FN5O3S | 50MG
SDP
Supplier:  Medchemexpress LLC HY12475950MG
Encompass_Preferred
CCR1 antagonist 9 is a potent, selective small-molecule antagonist of the chemokine receptor CCR1 intended for research use. It inhibits CCR1-mediated calcium flux with an IC50 of 6.8 nM, reduces CCR1-dependent chemotaxis in THP-1 cells, and shows measurable hERG activity.
  • Potent CCR1 inhibition with IC50 of 6.8 nM in calcium flux assay.
  • Inhibits CCR1 chemotaxis of THP-1 cells with IC50 of 28 nM.
  • Displays hERG activity with IC50 of 30 μM.
  • High purity (99.7%) and chemical formula C20H16FN5O3S.
  • Molecular weight 425.44 g/mol.
  • Available in multiple solid and solution pack sizes with COA and SDS documentation.
Catalog No. 50-002-35812