Learn More
Medchemexpress LLC 1-(4-fluorophenyl)-N-((2-(methylsulfonyl)pyridin-4-yl)methyl)-1H-pyrazolo[3,4-c]pyridine-4-carboxamide | 1220026-26-5 | 99.7% | 425.44 g·mol⁻¹ | C20H16FN5O3S | 10MG

Supplier: Medchemexpress LLC HY12475910MG
CCR1 antagonist 9 is a potent, selective small-molecule antagonist of the CC chemokine receptor 1 (CCR1) intended for laboratory research. It inhibits CCR1-mediated calcium flux with an IC50 of 6.8 nM, blocks CCR1 chemotaxis of THP-1 cells (IC50 28 nM), and shows hERG inhibition at higher concentrations (IC50 30 μM).
- Potent CCR1 antagonist with low-nanomolar activity in calcium flux assay.
- Demonstrated inhibition of CCR1 chemotaxis in THP-1 cells.
- High reported purity (≈99.7%).
- Chemical formula C20H16FN5O3S and molecular weight ~425.44 g·mol⁻¹.
- Typically supplied as a solid or as DMSO solutions for in vitro studies.
- Storage recommendations for powder and solutions to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.