Learn More
Medchemexpress LLC 1-(4-fluorophenyl)-N-[3-fluoro-4-(3-phenylpyrrolo[2,3-b]pyridin-4-yloxy)phenyl]-2,3-dimethyl-5-oxo | 1221713-92-3 | MFCD29905447 | 98.0% | 551.54 | C31H23F2N5O3 | 100 MG
SDP
Catalog No. 5000214070
Encompass_Preferred
NPS-1034 is a small-molecule dual inhibitor of the receptor tyrosine kinases AXL and MET intended for preclinical research. It exhibits low-nanomolar biochemical potency and is used to probe AXL/MET signaling and evaluate anti-tumor efficacy in cell-based and in vivo models.
  • Dual AXL and MET inhibition with low-nanomolar potency (AXL ≈10.3 nM; MET ≈48 nM).
  • Suitable for cell-based assays and in vivo oncology studies.
  • High chemical purity (>98.0%).
  • Molecular formula C31H23F2N5O3 and molecular weight 551.54 g/mol.
  • Available in multiple pack sizes and solution formats for experimental flexibility.
Catalog No. 50-002-14070 Supplier Medchemexpress LLC Supplier No. HY100509100MG
NPS-1034 is a small-molecule dual inhibitor of the receptor tyrosine kinases AXL and MET intended for preclinical research. It exhibits low-nanomolar biochemical potency and is used to probe AXL/MET signaling and evaluate anti-tumor efficacy in cell-based and in vivo models.
  • Dual AXL and MET inhibition with low-nanomolar potency (AXL ≈10.3 nM; MET ≈48 nM).
  • Suitable for cell-based assays and in vivo oncology studies.
  • High chemical purity (>98.0%).
  • Molecular formula C31H23F2N5O3 and molecular weight 551.54 g/mol.
  • Available in multiple pack sizes and solution formats for experimental flexibility.