Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)piperazine, 98%, Thermo Scientific™

Catalog No. AAA1454114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1454114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1454114 Supplier Thermo Scientific Chemicals Supplier No. A1454114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-(4-Fluorophenyl)piperazine was used in the synthesis of N,N-disubstituted piperazine. It is a major metabolite of niaparazine, a sedative, a hypnotic drug.

Solubility
Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from air, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2252-63-3
Molecular Formula C10H13FN2
Molecular Weight (g/mol) 180.226
MDL Number MFCD00005960
InChI Key AVJKDKWRVSSJPK-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl piperazine, 4-fluorophenylpiperazine, pfpp, 4-fluoro-phenylpiperazine, n-4-fluorophenyl piperazine, para-fluorophenylpiperazine, unii-vml61be244, 1-p-fluorophenyl piperazine, piperazine, 1-4-fluorophenyl, 4-fluorophenyl piperazine
PubChem CID 75260
IUPAC Name 1-(4-fluorophenyl)piperazine
SMILES C1CN(CCN1)C2=CC=C(C=C2)F

Specifications

Melting Point 30°C to 33°C
Boiling Point 120°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.56
Quantity 25 g
UN Number UN2923
Beilstein 610723
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 180.23
Percent Purity 98%
Chemical Name or Material 1-(4-Fluorophenyl)piperazine
Show More Show Less
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H301-H314
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 218-846-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.