Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(4-fluorophenyl)piperazine hydrochloride | 64090-19-3 | MFCD00012765 | 99.6% | 253.15 g·mol⁻¹ | C10H15Cl2FN2 | 25 G
SDP

Catalog No. 5000304198
Encompass_Preferred

1-(4-Fluorophenyl)piperazine hydrochloride is the hydrochloride salt of a substituted phenylpiperazine used as a research reagent and intermediate for synthesis. It is supplied as a solid hydrochloride with supporting documentation for laboratory use (not for human consumption).

  • High purity (99.59%) suitable for analytical and synthetic applications.
  • Hydrochloride salt in solid form for stable handling and storage.
  • Molecular weight 253.15 g·mol⁻¹; formula C10H15Cl2FN2.
  • Commonly used as a building block in medicinal chemistry and pharmacology research.
  • Accompanied by COA and SDS for quality and safety information.

Catalog No. 50-003-04198 Supplier Medchemexpress LLC Supplier No. 25GHYW016474A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(4-Fluorophenyl)piperazine hydrochloride is the hydrochloride salt of a substituted phenylpiperazine used as a research reagent and intermediate for synthesis. It is supplied as a solid hydrochloride with supporting documentation for laboratory use (not for human consumption).

  • High purity (99.59%) suitable for analytical and synthetic applications.
  • Hydrochloride salt in solid form for stable handling and storage.
  • Molecular weight 253.15 g·mol⁻¹; formula C10H15Cl2FN2.
  • Commonly used as a building block in medicinal chemistry and pharmacology research.
  • Accompanied by COA and SDS for quality and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.