Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Formylphenyl)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. F06795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06795G 5 g
F06791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06795G Supplier TCI America Supplier No. F06795G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51980-54-2
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00015469
InChI Key DATXHLPRESKQJK-UHFFFAOYSA-N
Synonym 4-1-pyrrolidinyl benzaldehyde, 4-pyrrolidin-1-yl benzaldehyde, 4-1-pyrrolidino benzaldehyde, 1-4-formylphenyl pyrrolidine, benzaldehyde, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-benzaldehyde, 4-pyrrolidinobenzaldehyde, 4-pyrrolidinylbenzaldehyde, 4-1-pyrrolidino-benzaldehyde, 4-pyrrolizinobenzaldehyde
PubChem CID 104037
IUPAC Name 4-pyrrolidin-1-ylbenzaldehyde
SMILES C1CCN(C1)C2=CC=C(C=C2)C=O

Specifications

Melting Point 85°C
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Formylphenyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.