Learn More
Chemglass Life Sciences 1/4 Hose Barb Genderless AseptiQuik S Connector, High Temperature
SDP
Catalog No. 5000980604
Encompass
Material: Polycarbonate, USP Class VI, ADCF. Material Finish: Molded White. Pressure Range: Up to 60 psi, 4. 1 bar. Color: White with white tab covers. Mounting Option: Free Floating. Sterilization: Autoclave and Gamma Radiation Compatible. Temperature Range: 39 - 104 ° F (4 - 40 ° C). Seal Material: Silcone, Platinum-cured, USP Class VI, ADCF. Valve Option: Non-Valved.
Catalog No. 50-009-80604 Supplier Chemglass Life Sciences Supplier No. CLS91309004HT
Material: Polycarbonate, USP Class VI, ADCF. Material Finish: Molded White. Pressure Range: Up to 60 psi, 4. 1 bar. Color: White with white tab covers. Mounting Option: Free Floating. Sterilization: Autoclave and Gamma Radiation Compatible. Temperature Range: 39 - 104 ° F (4 - 40 ° C). Seal Material: Silcone, Platinum-cured, USP Class VI, ADCF. Valve Option: Non-Valved.