Learn More
Chemglass Life Sciences 1/4" Hose Barb Non-Valved In-Line Coupling Body, Polysulfone, CPC MPC17004T39
SDP
Catalog No. 5000980705
Encompass
MPC Series couplings add ease of use and security to your most critical fluid handling applications. Choose from a full line of connectors and configurations, including pressure sealing caps and plugs, in sizes to fit 1/8", 1/4" and 3/8" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks. Features: Ergonomic thumb latchUSP Class VI materialsSterilizable by autoclave, Et0, e-beam or gammaParting line-free hose barbBSE/TSE free materials   Benefits: Easy to operate – even with gloved handsMeet biocompatibility requirementsReusable, yet economical enough to allow disposabilityEliminates potential leak pathMeet ADCF requirements.
Catalog No. 50-009-80705 Supplier Chemglass Life Sciences Supplier No. CLS91404039
MPC Series couplings add ease of use and security to your most critical fluid handling applications. Choose from a full line of connectors and configurations, including pressure sealing caps and plugs, in sizes to fit 1/8", 1/4" and 3/8" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks. Features: Ergonomic thumb latchUSP Class VI materialsSterilizable by autoclave, Et0, e-beam or gammaParting line-free hose barbBSE/TSE free materials   Benefits: Easy to operate – even with gloved handsMeet biocompatibility requirementsReusable, yet economical enough to allow disposabilityEliminates potential leak pathMeet ADCF requirements.