Learn More
New Jersey Fluid Systems LLC 1/4" HOSE QUICK CONNECT NPT
Catalog No. NC0575878
Encompass
316 Stainless Steel Swagelok QC4 in. series Quick Connect Shut-Off Stem; 1/4 in. female NPT end; Viton o-ring; 3,000psi @100F
Catalog No. NC0575878 Supplier New Jersey Fluid Systems LLC Supplier No. SSQC4D4PF
316 Stainless Steel Swagelok QC4 in. series Quick Connect Shut-Off Stem; 1/4 in. female NPT end; Viton o-ring; 3,000psi @100F