Learn More
Ampco 1/4 in. to 1/2 in. Ratchet Adapter
SDP
Catalog No. 19174625 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
15/16 in.
Catalog No. Length (English)
19-174-625 15/16 in.
1 options
Catalog No. 19-174-625 Supplier Ampco Supplier No. W143RA
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion resistant ratchet adapter. Frequently purchased with Ampco ratchet wrench.
  • Frequently purchased with ratchet wrench.
  • Made in the USA. HS Code: 8207.90.
  • See image for additional information regarding measurements.
  • End 1 size (a1): 1/4 in.
  • End 2 size (a2): 1/2 in.
  • Length: 15/16 in.
Specifications
Product Type Ratchet Adapter
Length (English) 15/16 in.
Quantity 1 Ea.