Learn More
New Jersey Fluid Systems LLC 1/4 IN FNPT HEX COUPLING
Catalog No. NC9317982
Encompass
316 Stainless Steel Swagelok Pipe Fitting Hex Coupling. 1/4 in. Female NPT
Catalog No. NC9317982 Supplier New Jersey Fluid Systems LLC Supplier No. SS4HCG
316 Stainless Steel Swagelok Pipe Fitting Hex Coupling. 1/4 in. Female NPT