Learn More
Waters Corp PTFE Tubing, 1/4 in O.D., 0.19 in I.D., 10 ft Length
Catalog No. 50785260 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters tubing and fittings are carefully designed to provide leak-free connections, with low dead volume, and minimal band spreading. For best performance, these components must be clean, and designed to work together. The geometry of the ferrule and the conical port that it is used with must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths and diameters are available to provide the flexibility you need when you set up and maintain your system.
  • Provides leak-free connections
  • Features low dead volume
  • Designed for minimal band spreading
  • Components are clean and designed to work together for best performance
  • Geometry of ferrule and conical port must match
  • Diameter of tube and fittings must match
  • Variety of materials, lengths, and diameters are available for system setup and maintenance
Catalog No. 50-785-260 Supplier Waters Corp Supplier No. WAT026810
Waters tubing and fittings are carefully designed to provide leak-free connections, with low dead volume, and minimal band spreading. For best performance, these components must be clean, and designed to work together. The geometry of the ferrule and the conical port that it is used with must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths and diameters are available to provide the flexibility you need when you set up and maintain your system.
  • Provides leak-free connections
  • Features low dead volume
  • Designed for minimal band spreading
  • Components are clean and designed to work together for best performance
  • Geometry of ferrule and conical port must match
  • Diameter of tube and fittings must match
  • Variety of materials, lengths, and diameters are available for system setup and maintenance