Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Iodophenyl)hydrazine 95%, Thermo Scientific™

Catalog No. RF01374DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RF01374DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF01374DA Supplier Thermo Scientific Chemicals Supplier No. RF01374DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13116-27-3
Molecular Formula C6H7IN2
Molecular Weight (g/mol) 234.04
InChI Key IQMLCMKMSBMMGR-UHFFFAOYSA-N
Synonym 4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine
PubChem CID 704591
IUPAC Name (4-iodophenyl)hydrazine
SMILES C1=CC(=CC=C1NN)I

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula IC6H4NHNH2
Quantity 1 g
Formula Weight 234.04
Percent Purity 95%
Chemical Name or Material 1-(4-Iodophenyl)hydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.