Learn More
Medchemexpress LLC 1-(4-methoxybenzyl)-5-trifluoromethoxyisatin | 1160247-92-6 | 99.9% | 25 MG
SDP
Catalog No. 5000234748
Encompass_Preferred
VU 0238429 is a research-grade positive allosteric modulator of muscarinic acetylcholine receptor subtype 5 (mAChR5). In vitro it shows an EC50 of 1.16 μM and >30-fold selectivity versus M1 and M3, with no potentiation of M2 or M4. Supplied as a high-purity solid or as DMSO solutions, it is intended for laboratory research use.
  • Positive allosteric modulator of mAChR5.
  • In vitro potency EC50 1.16 μM.
  • Greater than 30-fold selectivity versus M1 and M3.
  • High purity (≈99.9%).
  • Available as solid or 10 mM solution in DMSO.
  • Recommended storage: powder -20°C; in solvent -80°C for long-term.
Catalog No. 50-002-34748 Supplier Medchemexpress LLC Supplier No. HY1215725MG
VU 0238429 is a research-grade positive allosteric modulator of muscarinic acetylcholine receptor subtype 5 (mAChR5). In vitro it shows an EC50 of 1.16 μM and >30-fold selectivity versus M1 and M3, with no potentiation of M2 or M4. Supplied as a high-purity solid or as DMSO solutions, it is intended for laboratory research use.
  • Positive allosteric modulator of mAChR5.
  • In vitro potency EC50 1.16 μM.
  • Greater than 30-fold selectivity versus M1 and M3.
  • High purity (≈99.9%).
  • Available as solid or 10 mM solution in DMSO.
  • Recommended storage: powder -20°C; in solvent -80°C for long-term.