Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(4-methoxyphenyl)-3-methyl-1H-pyrazol-5(4H)-one | 60798-06-3 | MFCD01361012 | 99.5% | 204.22 g/mol | C11H12N2O2 | 250 MG
SDP

Catalog No. 5000209295
Encompass_Preferred

  • reported antiprion activity in ScN2a and F3 cell assays (IC50 ~13 nM and 25 nM)
  • high chemical purity suitable for biological assays
  • well-defined molecular identity (CAS 60798-06-3, C11H12N2O2)
  • commonly used in small-molecule screening and neurodegeneration research

Catalog No. 50-002-09295 Supplier Medchemexpress LLC Supplier No. 250MGHYW081348
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • reported antiprion activity in ScN2a and F3 cell assays (IC50 ~13 nM and 25 nM)
  • high chemical purity suitable for biological assays
  • well-defined molecular identity (CAS 60798-06-3, C11H12N2O2)
  • commonly used in small-molecule screening and neurodegeneration research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.