Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Methoxyphenyl)imidazoline-2-thione, 98%, Thermo Scientific™

Catalog No. p-7045849
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1221603 1 g
AAL1221606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1221603 Supplier Thermo Scientific Chemicals Supplier No. L1221603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17452-14-1
Molecular Formula C10H10N2OS
Molecular Weight (g/mol) 206.26
MDL Number MFCD00041376
InChI Key LIEBCNQPMNARMP-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl imidazoline-2-thione, 1-4-methoxyphenyl-1h-imidazole-2 3h-thione, 1-4-methoxyphenyl-1h-imidazole-2-thiol, 1-4-methoxyphenyl imidazole-2-thiol, 1-4-methoxyphenyl-3h-imidazole-2-thione, chembl83498, 1-4-methoxyphenyl-2-mercaptoimidazole, 1-4-methoxyphenyl-4-imidazoline-2-thione, 3-4-methoxyphenyl-1h-imidazole-2-thione, 1-4-methoxy-phenyl-1,3-dihydro-imidazole-2-thione
PubChem CID 2759656
IUPAC Name 1-(4-methoxyphenyl)-2,3-dihydro-1H-imidazole-2-thione
SMILES COC1=CC=C(C=C1)N1C=CNC1=S

Specifications

Melting Point 216°C to 220°C
Quantity 1 g
Beilstein 155898
Formula Weight 206.27
Percent Purity 98%
Chemical Name or Material 1-(4-Methoxyphenyl)imidazoline-2-thione
EINECSNumber 674-113-4
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.