Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[(4-methoxyphenyl)methyl]-5-(trifluoromethoxy)indole-2,3-dione | 1160247-92-6 | 99.9% | 10 MG
SDP

Catalog No. 5000231536
Encompass_Preferred

VU 0238429 is a selective positive allosteric modulator of the muscarinic acetylcholine receptor subtype M5 (mAChR5), with an EC50 of approximately 1.16 μM. It is used as a research tool to probe M5-mediated GPCR signaling and neuronal pharmacology.

  • Molecular formula: C17H12F3NO4.
  • Molecular weight: 351.28 g/mol.
  • CAS number: 1160247-92-6.
  • Purity: ~99.9% by HPLC.
  • Solubility: ≥150 mg/mL in DMSO; ≥2.5 mg/mL in 10% DMSO/40% PEG300/5% Tween-80/45% saline for in vivo formulations.
  • Target: muscarinic acetylcholine receptor M5 (mAChR5).
  • EC50: ~1.16 μM at M5; >30 μM at M1 and M3 indicating selectivity.
  • Storage: powder -20°C (long term), 4°C (short term); solutions -80°C (up to 6 months).
  • Quantity: supplied as a 10 mg solid.

Catalog No. 50-002-31536 Supplier Medchemexpress LLC Supplier No. HY1215710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU 0238429 is a selective positive allosteric modulator of the muscarinic acetylcholine receptor subtype M5 (mAChR5), with an EC50 of approximately 1.16 μM. It is used as a research tool to probe M5-mediated GPCR signaling and neuronal pharmacology.

  • Molecular formula: C17H12F3NO4.
  • Molecular weight: 351.28 g/mol.
  • CAS number: 1160247-92-6.
  • Purity: ~99.9% by HPLC.
  • Solubility: ≥150 mg/mL in DMSO; ≥2.5 mg/mL in 10% DMSO/40% PEG300/5% Tween-80/45% saline for in vivo formulations.
  • Target: muscarinic acetylcholine receptor M5 (mAChR5).
  • EC50: ~1.16 μM at M5; >30 μM at M1 and M3 indicating selectivity.
  • Storage: powder -20°C (long term), 4°C (short term); solutions -80°C (up to 6 months).
  • Quantity: supplied as a 10 mg solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.