Learn More
Medchemexpress LLC 1-[(4-methoxyphenyl)methyl]-5-(trifluoromethoxy)indole-2,3-dione | 1160247-92-6 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1215750MG
Encompass_Preferred
VU 0238429 is a small-molecule research compound that acts as a positive allosteric modulator of the muscarinic acetylcholine receptor subtype 5 (mAChR5, M5). It potentiates M5 with an EC50 of 1.16 μM, shows >30-fold selectivity versus M1 and M3, and is supplied as solid or as a 10 mM solution in DMSO at high purity for laboratory research.
  • Positive allosteric modulator of mAChR5 with EC50 1.16 μM.
  • Greater than 30-fold selectivity versus M1 and M3; no M2 or M4 potentiation.
  • Available as solid (multiple sizes) and 10 mM solution in DMSO.
  • High purity (listed around 99.9%).
  • Intended for laboratory research use only; not for human use.
Catalog No. 50-002-31236