Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Methoxyphenyl)piperazine, 97%

Catalog No. AAH55791MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55791MD 250 mg
AAH5579103 1 g
AAH5579106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55791MD Supplier Thermo Scientific Chemicals Supplier No. H55791MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 38212-30-5 | C11H16N2O | 192.26 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
soluble in water, methanol and toluene

Notes
Air Sensitive. Store under nitrogen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38212-30-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.26
MDL Number MFCD00040736
InChI Key MRDGZSKYFPGAKP-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl piperazine, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg, 4-methoxyphenylpiperazine, 1-4-anisyl piperazine, chembl59517, 1-4-methoxyphenyl-piperazine, 1-4-methoxy-phenyl-piperazine
PubChem CID 269722
IUPAC Name 1-(4-methoxyphenyl)piperazine
SMILES COC1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 42°C to 47°C
Quantity 250 mg
Beilstein 156736
Sensitivity Air Sensitive
Solubility Information soluble in water,methanol and toluene
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material 1-(4-Methoxyphenyl)piperazine
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 253-829-7
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.