Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(4-Methoxyphenyl)piperazine hydrochloride | 38869-47-5 | 99.2% | 265.18 | 100 G
SDP

Catalog No. 5000452578
Encompass_Preferred

1-(4-Methoxyphenyl)piperazine hydrochloride is a piperazine derivative exhibiting neural activity. It is used in neurological disease research as a drug derivative.

  • CAS number: 38869-47-5
  • Molecular formula: C11H18Cl2N2O
  • Molecular weight: 265.18
  • Purity: 99.2%
  • Appearance: Off-white to light brown solid
  • Recommended storage: Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-52578 Supplier Medchemexpress LLC Supplier No. HYW011726100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(4-Methoxyphenyl)piperazine hydrochloride is a piperazine derivative exhibiting neural activity. It is used in neurological disease research as a drug derivative.

  • CAS number: 38869-47-5
  • Molecular formula: C11H18Cl2N2O
  • Molecular weight: 265.18
  • Purity: 99.2%
  • Appearance: Off-white to light brown solid
  • Recommended storage: Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.