Learn More
Medchemexpress LLC 1,4-naphthoquinone | 130-15-4 | MFCD00001676 | 99.5% | 158.16 g·mol⁻¹ | C10H6O2 | 1 ML
SDP
Catalog No. 5000302436
Encompass_Preferred
1,4-Naphthoquinone is a quinone-family small molecule used in biochemical and pharmacological research, including studies of monoamine oxidase inhibition, DNA/topoisomerase interactions, and oxidative stress. This item is supplied as a 10 mM solution in DMSO in a 1 mL vial for ready use in assays and screening.
Catalog No. 50-003-02436 Supplier Medchemexpress LLC Supplier No. HYW01549010MM/1ML
1,4-Naphthoquinone is a quinone-family small molecule used in biochemical and pharmacological research, including studies of monoamine oxidase inhibition, DNA/topoisomerase interactions, and oxidative stress. This item is supplied as a 10 mM solution in DMSO in a 1 mL vial for ready use in assays and screening.