Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-naphthoquinone | 130-15-4 | MFCD00001676 | 99.5% | 158.16 g/mol | C10H6O2 | 100 G
SDP

Catalog No. 5000303756
Encompass_Preferred

1,4-Naphthoquinone (CAS 130-15-4) is an organic quinone used as a biochemical research reagent. It is reported to act as an inhibitor in studies of DNA/RNA synthesis, NF-κB signaling, and monoamine oxidases, and is supplied as a stable powder for laboratory use.

  • High purity (reported ~99.5%).
  • Molecular formula C10H6O2.
  • Molecular weight 158.16 g/mol.
  • Used as an inhibitor in biochemical and cell-based assays.
  • Supplied as a stable powder with supplier storage and handling guidance.

Catalog No. 50-003-03756 Supplier Medchemexpress LLC Supplier No. HYW015490100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-Naphthoquinone (CAS 130-15-4) is an organic quinone used as a biochemical research reagent. It is reported to act as an inhibitor in studies of DNA/RNA synthesis, NF-κB signaling, and monoamine oxidases, and is supplied as a stable powder for laboratory use.

  • High purity (reported ~99.5%).
  • Molecular formula C10H6O2.
  • Molecular weight 158.16 g/mol.
  • Used as an inhibitor in biochemical and cell-based assays.
  • Supplied as a stable powder with supplier storage and handling guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.