Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-Naphthoquinone | 130-15-4 | MFCD00001676 | 99.5% | 158.16 g/mol | C10H6O2 | 500 G
SDP

Catalog No. 5000303758
Encompass_Preferred

1,4-Naphthoquinone (CAS 130-15-4) is a research-grade small-molecule quinone used in biochemical and pharmacological studies. It acts as an MAO-A/B inhibitor and is used in antibacterial, antitumor, and anti-inflammatory research. Supplied as a solid powder with documented purity and storage recommendations, it is appropriate for in vitro assays and synthesis applications.

  • High purity suitable for research applications.
  • Molecular formula C10H6O2; molecular weight 158.16 g/mol.
  • Available in solid form, including 500 g package sizes.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
  • Storage recommendations provided for powder and solutions.
  • Suitable for biochemical assays targeting monoamine oxidases and related studies.

Catalog No. 50-003-03758 Supplier Medchemexpress LLC Supplier No. HYW015490500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-Naphthoquinone (CAS 130-15-4) is a research-grade small-molecule quinone used in biochemical and pharmacological studies. It acts as an MAO-A/B inhibitor and is used in antibacterial, antitumor, and anti-inflammatory research. Supplied as a solid powder with documented purity and storage recommendations, it is appropriate for in vitro assays and synthesis applications.

  • High purity suitable for research applications.
  • Molecular formula C10H6O2; molecular weight 158.16 g/mol.
  • Available in solid form, including 500 g package sizes.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
  • Storage recommendations provided for powder and solutions.
  • Suitable for biochemical assays targeting monoamine oxidases and related studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.