Learn More
Medchemexpress LLC 1,4-naphthoquinone-d6 | 26473-08-5 | 99.2% | 164.19 | C10D6O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW015490S
Encompass_Preferred
1,4-Naphthoquinone-d6 is a deuterium-labeled analogue of 1,4-naphthoquinone used as an internal standard, tracer, and synthetic building block in analytical and preparative chemistry. It delivers a mass-shifted, chemically equivalent compound for accurate mass spectrometry, quantitation, and mechanistic studies.
  • Deuterium labeling (d6) for mass spectrometry and tracer studies.
  • High chemical purity (99.2%) suitable for analytical applications.
  • Molecular formula C10D6O2 and molecular weight 164.19.
  • Available in small pack sizes for research use.
  • Solid form with defined storage conditions to preserve stability.
Catalog No. 50-003-03420