Learn More
Medchemexpress LLC 1,4-naphthoquinone-d6 | 26473-08-5 | 99.2% | 164.19 g·mol⁻¹ | C10D6O2 | 1 MG
SDP
Catalog No. 5000303759
Encompass_Preferred
1,4-Naphthoquinone-d6 is the deuterium-labeled analogue of 1,4-naphthoquinone supplied as a high-purity research standard for analytical and biochemical applications, including isotope tracing and analytical method development.
  • deuterium-labeled (d6) analogue for isotope tracing and internal standard use.
  • high chemical purity: 99.2%.
  • molecular formula C10D6O2 and molecular weight 164.19 g·mol⁻¹.
  • supplied in small quantity packaging suitable for analytical applications.
  • accompanied by a datasheet and safety data sheet for laboratory use.
Catalog No. 50-003-03759 Supplier Medchemexpress LLC Supplier No. HYW015490S1MG
1,4-Naphthoquinone-d6 is the deuterium-labeled analogue of 1,4-naphthoquinone supplied as a high-purity research standard for analytical and biochemical applications, including isotope tracing and analytical method development.
  • deuterium-labeled (d6) analogue for isotope tracing and internal standard use.
  • high chemical purity: 99.2%.
  • molecular formula C10D6O2 and molecular weight 164.19 g·mol⁻¹.
  • supplied in small quantity packaging suitable for analytical applications.
  • accompanied by a datasheet and safety data sheet for laboratory use.