Learn More
Medchemexpress LLC 1,4-naphthoquinone-d6 | 26473-08-5 | 99.2% | 164.19 g/mol | C10D6O2 | 10 MG
SDP
Catalog No. 5000302534
Encompass_Preferred
1,4-Naphthoquinone-d6 is the deuterium-labeled analogue of 1,4-naphthoquinone supplied as a high-purity solid for use as a stable isotope standard, tracer, or analytical reference in mechanistic studies and mass spectrometry. The material is formulated and supplied with storage guidance to maintain isotopic and chemical integrity for laboratory use.
  • Deuterium labeled (d6) for isotope tracing and internal standard use.
  • High purity suitable for analytical applications (99.2%).
  • Solid, light yellow to yellow appearance for easy handling.
  • Supplied in small sizes for quality-controlled experiments.
  • Storage guidance supports long-term stability of powder and solutions.
Catalog No. 50-003-02534 Supplier Medchemexpress LLC Supplier No. HYW015490S10MG
1,4-Naphthoquinone-d6 is the deuterium-labeled analogue of 1,4-naphthoquinone supplied as a high-purity solid for use as a stable isotope standard, tracer, or analytical reference in mechanistic studies and mass spectrometry. The material is formulated and supplied with storage guidance to maintain isotopic and chemical integrity for laboratory use.
  • Deuterium labeled (d6) for isotope tracing and internal standard use.
  • High purity suitable for analytical applications (99.2%).
  • Solid, light yellow to yellow appearance for easy handling.
  • Supplied in small sizes for quality-controlled experiments.
  • Storage guidance supports long-term stability of powder and solutions.