Learn More
Medchemexpress LLC 1,4-naphthoquinone-d6 | 26473-08-5 | MFCD03428248 | 99.2% | 164.19 g/mol | C10D6O2 | 5 MG
SDP
Catalog No. 5000303757
Encompass_Preferred
1,4-Naphthoquinone-d6 is the deuterium-labeled analogue of 1,4-naphthoquinone (C10D6O2, MW 164.19 g/mol), provided as a stable-isotope research reagent and reference standard for analytical and mechanistic studies. It is intended for use in mass spectrometry, tracer experiments, and metabolic labeling to track naphthoquinone-related pathways. Purity is reported on the certificate of analysis.
  • Deuterium-labeled reference standard for analytical and tracer studies.
  • Molecular formula C10D6O2 and molecular weight 164.19 g/mol.
  • High isotopic enrichment and reported purity of 99.2%.
  • Supplied as a small-quantity solid suitable for mass spectrometry and mechanistic experiments.
  • Used for metabolic labeling, quantitation, and pathway analysis in research applications.
Catalog No. 50-003-03757 Supplier Medchemexpress LLC Supplier No. HYW015490S5MG
1,4-Naphthoquinone-d6 is the deuterium-labeled analogue of 1,4-naphthoquinone (C10D6O2, MW 164.19 g/mol), provided as a stable-isotope research reagent and reference standard for analytical and mechanistic studies. It is intended for use in mass spectrometry, tracer experiments, and metabolic labeling to track naphthoquinone-related pathways. Purity is reported on the certificate of analysis.
  • Deuterium-labeled reference standard for analytical and tracer studies.
  • Molecular formula C10D6O2 and molecular weight 164.19 g/mol.
  • High isotopic enrichment and reported purity of 99.2%.
  • Supplied as a small-quantity solid suitable for mass spectrometry and mechanistic experiments.
  • Used for metabolic labeling, quantitation, and pathway analysis in research applications.