Learn More
Medchemexpress LLC 1,4-naphthoquinone (standard) | 130-15-4 | MFCD00001676 | 99.5% | 158.16 g/mol | C10H6O2 | 50 G
SDP
Catalog No. 5000303639
Encompass_Preferred
1,4-Naphthoquinone is a small-molecule quinone used as a research reagent in biochemical and pharmacological studies. It is reported to inhibit monoamine oxidases and is commonly used to probe DNA/RNA synthesis, oxidative stress, and antimicrobial or antitumor mechanisms.
  • Used as a biochemical probe for MAO-A/B and related enzyme studies.
  • Supports studies of DNA/RNA synthesis and oxidative damage pathways.
  • Available in multi-gram pack sizes suitable for synthesis and assays.
  • High purity suitable for research applications.
  • Documented safety and analytical data provided (SDS, COA, HPLC).
Catalog No. 50-003-03639 Supplier Medchemexpress LLC Supplier No. HYW01549050G
1,4-Naphthoquinone is a small-molecule quinone used as a research reagent in biochemical and pharmacological studies. It is reported to inhibit monoamine oxidases and is commonly used to probe DNA/RNA synthesis, oxidative stress, and antimicrobial or antitumor mechanisms.
  • Used as a biochemical probe for MAO-A/B and related enzyme studies.
  • Supports studies of DNA/RNA synthesis and oxidative damage pathways.
  • Available in multi-gram pack sizes suitable for synthesis and assays.
  • High purity suitable for research applications.
  • Documented safety and analytical data provided (SDS, COA, HPLC).