Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Nitrobenzyl)-1,2,4-triazole 98.0+%, TCI America™
SDP

Catalog No. N10075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10075G Supplier TCI America Supplier No. N10075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 119192-09-5
Melting Point 102°C
Molecular Formula C9H8N4O2
Quantity 5 g
MDL Number MFCD03840588
InChI Key NVRYCUYVBBCXHT-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CN2C=NC=N2)[N+](=O)[O-]
IUPAC Name 1-[(4-nitrophenyl)methyl]-1,2,4-triazole
Molecular Weight (g/mol) 204.189
PubChem CID 6482000
Formula Weight 204.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Nitrobenzyl)-1,2,4-triazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.