Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(((4-Nitrophenoxy)carbonyl)oxy)ethyl isobutyrate | 194995-47-6 | 99.9% | 297.26 | 250 MG
SDP

Catalog No. 5000402245
Encompass_Preferred

1-(((4-Nitrophenoxy)carbonyl)oxy)ethyl isobutyrate is a drug intermediate for the synthesis of various active compounds. This product is for research use only and not sold to patients.

  • Available in solid appearance
  • Color: white to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, stored under nitrogen
  • In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Catalog No. 50-004-02245 Supplier Medchemexpress LLC Supplier No. 250MGHY21206
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-(((4-Nitrophenoxy)carbonyl)oxy)ethyl isobutyrate is a drug intermediate for the synthesis of various active compounds. This product is for research use only and not sold to patients.

  • Available in solid appearance
  • Color: white to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, stored under nitrogen
  • In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.