Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Nitrophenyl)piperidine, 97%, Thermo Scientific™

Catalog No. AAB2039914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2039914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2039914 Supplier Thermo Scientific Chemicals Supplier No. B2039914

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6574-15-8
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD00023662
InChI Key SGPLAXFUDTWHRS-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl piperidine, 1-p-nitrophenyl piperidine, n-4-nitrophenyl piperidine, 1-4-nitro-phenyl-piperidine, piperidine, 1-4-nitrophenyl, n-p-nitrophenyl piperidine, 4-nitrophenyl piperidine, pubchem9438, maybridge1_000682, acmc-20a12d
PubChem CID 81049
IUPAC Name 1-(4-nitrophenyl)piperidine
SMILES C1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 102°C to 105°C
Quantity 25 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 1-(4-Nitrophenyl)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-492-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.