Learn More
Ace Glass, Inc. PTFE insert, 1/4" NPT, #7 compression fitting6.5 to 7mm OD tubing
SDP
Catalog No. 50983474
Encompass
Compression fitting PTFE inserts for 1/4NPT to #7 Ace-Thred. Fits 6.5 to 7mm O.D. tubing. For use with ACE PTFE reaction vessel heads.
Catalog No. 50-983-474 Supplier Ace Glass, Inc. Supplier No. 1287103
Compression fitting PTFE inserts for 1/4NPT to #7 Ace-Thred. Fits 6.5 to 7mm O.D. tubing. For use with ACE PTFE reaction vessel heads.