Learn More
MSC CPC Colder Products PMC1004 1/8" Nominal Flow, 1/4 NPT Thread, Female, Inline Threaded-Female Socket 120 Max psi, -40 to 180°F, Acetal Coupling, Buna-N O-Ring
Catalog No. NC1725896
Encompass
CPC Colder Products PMC1004 1/8" Nominal Flow, 1/4 NPT Thread, Female, Inline Threaded-Female Socket 120 Max psi, -40 to 180°F, Acetal Coupling, Buna-N O-Ring
Catalog No. NC1725896 Supplier MSC Supplier No. 48607063
CPC Colder Products PMC1004 1/8" Nominal Flow, 1/4 NPT Thread, Female, Inline Threaded-Female Socket 120 Max psi, -40 to 180°F, Acetal Coupling, Buna-N O-Ring