Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Octadiene (cis- and trans- mixture), TCI America™
SDP

Catalog No. O01701ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01701ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01701ML Supplier TCI America Supplier No. O01701ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5675-25-2
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00027235
InChI Key HYBLFDUGSBOMPI-FPLPWBNLSA-N
PubChem CID 5462702
IUPAC Name (4Z)-octa-1,4-diene
SMILES CCCC=CCC=C

Specifications

Color Yellow
Quantity 1 mL
UN Number 2309
Formula Weight 110.20
Physical Form Liquid
Chemical Name or Material 1,4-Octadiene (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.