Learn More
Medchemexpress LLC 1,4-Oxazepin-5(2H)-one, 4-[3-[[2-[[2-ethyl-4-(4-methyl-1-piperazinyl)phenyl)amino]-5-(trifluoromethyl)-4-pyrimidinyl]amino]propyl]tetrahydro- | 2543673-19-2 | 99.7% | 535.60 | 500 MG
SDP
Catalog No. 5000394407
Encompass_Preferred
DCC-3116 is an orally active ULK1/2 inhibitor. It inhibits autophagy in lung cancer cells by targeting KRASG12C signaling, which in turn inhibits the proliferation of lung cancer cells and exhibits anti-cancer effects.
  • Orally active ULK1/2 inhibitor.
  • Inhibits autophagy in lung cancer cells.
  • Inhibits KRASG12C signaling.
  • Exerts anti-cancer effects.
  • Available in various quantities.
Catalog No. 50-003-94407 Supplier Medchemexpress LLC Supplier No. HY160699500MG
DCC-3116 is an orally active ULK1/2 inhibitor. It inhibits autophagy in lung cancer cells by targeting KRASG12C signaling, which in turn inhibits the proliferation of lung cancer cells and exhibits anti-cancer effects.
  • Orally active ULK1/2 inhibitor.
  • Inhibits autophagy in lung cancer cells.
  • Inhibits KRASG12C signaling.
  • Exerts anti-cancer effects.
  • Available in various quantities.