Learn More
Medchemexpress LLC 1,4-PBIT (dihydrobromide) | 157254-60-9 | 98.0% | 444.25 g·mol⁻¹ | C12H20Br2N4S2 | 5 MG
SDP
Catalog No. 5000222005
Encompass_Preferred
1,4-PBIT (dihydrobromide) is a research-grade small-molecule inhibitor of nitric oxide synthases provided as the dihydrobromide salt (CAS 157254-60-9). Supplied in small-mass vials for biochemical and pharmacological studies, the compound is characterized by analytical documentation and high purity suitable for in vitro research.
  • Potent inhibitor of iNOS, eNOS, and nNOS isoforms.
  • High purity for reliable experimental results.
  • Available in small-mass vials suitable for medicinal chemistry applications.
  • Stable dihydrobromide salt form for consistent handling.
  • Documented analytical data and certificate of analysis available.
Catalog No. 50-002-22005 Supplier Medchemexpress LLC Supplier No. HY1352245MG
1,4-PBIT (dihydrobromide) is a research-grade small-molecule inhibitor of nitric oxide synthases provided as the dihydrobromide salt (CAS 157254-60-9). Supplied in small-mass vials for biochemical and pharmacological studies, the compound is characterized by analytical documentation and high purity suitable for in vitro research.
  • Potent inhibitor of iNOS, eNOS, and nNOS isoforms.
  • High purity for reliable experimental results.
  • Available in small-mass vials suitable for medicinal chemistry applications.
  • Stable dihydrobromide salt form for consistent handling.
  • Documented analytical data and certificate of analysis available.