Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, dihydrochloride | 56100-42-6 | 98.4% | 472.88 | 1 ML
SDP

Catalog No. 5000352248
Encompass_Preferred

l-Atabrine dihydrochloride is a less active enantiomer of quinacrine that displays antiprion activity. It is a light yellow to yellow solid with a purity of 98.35%.

  • Displays antiprion activity.
  • Intended for research use only.
  • Soluble in H₂O and DMSO.
  • Ships at room temperature.
  • Store at 4°C, sealed, away from moisture.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-003-52248 Supplier Medchemexpress LLC Supplier No. HY13735C10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

l-Atabrine dihydrochloride is a less active enantiomer of quinacrine that displays antiprion activity. It is a light yellow to yellow solid with a purity of 98.35%.

  • Displays antiprion activity.
  • Intended for research use only.
  • Soluble in H₂O and DMSO.
  • Ships at room temperature.
  • Store at 4°C, sealed, away from moisture.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.