Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Phenylene diisocyanate

Catalog No. 262242100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-224-2100G 100 g
26-224-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-224-2100G Supplier Sigma Aldrich Supplier No. 262242100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Phenylene diisocyanate

Specifications

CAS 104-49-4
Melting Point 96°C to 99°C (lit.)
Boiling Point 260°C (lit.)
Molecular Formula C8H4N2O2
Linear Formula C6H4(NCO)2
MDL Number MFCD00002025
Quantity 100 g
Molecular Weight (g/mol) 160.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.