Learn More
Bioworld 1,4-Phenylene Diisothiocyanate, 1 g
Supplier:  Bioworld 212100001
Encompass_Preferred
1, 4-Phenylene disothiocyanateM.W.= 192.26Solid phase protein sequencingReference: 1.) Laursen, R.A., et al. (1972) FEBS Lett. 21, 67.
Catalog No. 50-198-570