Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Phenylenediboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P13581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13581G Supplier TCI America Supplier No. P13581G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4612-26-4
Molecular Formula C6H8B2O4
Molecular Weight (g/mol) 165.746
MDL Number MFCD00236018
InChI Key BODYVHJTUHHINQ-UHFFFAOYSA-N
Synonym 1,4-benzenediboronic acid, 1,4-phenylenediboronic acid, 1,4-phenylenebisboronic acid, benzene-1,4-diboronic acid, 4-boronophenyl boronic acid, p-benzenediboronic acid, p-phenylenediboronic acid, 1,4-phenylenebis boronic acid, 1,4 phenyl diboronic acid, boronic acid, 1,4-phenylenebis
PubChem CID 230478
IUPAC Name (4-boronophenyl)boronic acid
SMILES B(C1=CC=C(C=C1)B(O)O)(O)O

Specifications

Quantity 1 g
Formula Weight 165.75
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylenediboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.