Learn More
Medchemexpress LLC 1-(4-Pip-Ph)-DHP-dione | 2446913-96-6 | 274.32 | C14H18N4O2 | 50mg
SDP
Catalog No. 5000270223
Encompass_Preferred
1-(4-Pip-Ph)-DHP-dione is an E3 ligase ligand 1-(4-Pip-Ph)-DHP-dione can be used for synthesis of PROTAC HPK1 Degrader-2 (HY-162544)[1]
Catalog No. 50-002-70223 Supplier Medchemexpress LLC Supplier No. HYW76130050MG
1-(4-Pip-Ph)-DHP-dione is an E3 ligase ligand 1-(4-Pip-Ph)-DHP-dione can be used for synthesis of PROTAC HPK1 Degrader-2 (HY-162544)[1]