Learn More
Medchemexpress LLC 1,4-Piperazinedipropanesulfonic acid, β1,4-dihydroxy- | 68189-43-5 | ≥95.0% | 25 G
SDP
Catalog No. 5000414315
Encompass_Preferred
POPSO is a zwitterionic buffer that increases osmolality and demonstrates significant inhibition of anion uniport. It inhibits chloride uniport with an IC50 value of 24 mM. POPSO also enhances copper uptake and toxicity in alga and impairs the mitochondrial inner membrane. The working pH range for POPSO sesquisodium salt is 7.2-8.5.
  • Zwitterionic buffer
  • Increases osmolality
  • Marked inhibition of anion uniport
  • Inhibits chloride uniport with an IC50 value of 24 mM
  • Enhances copper uptake and toxicity in alga
  • Impairs mitochondrial inner membrane
  • Working pH range (for sesquisodium salt): 7.2-8.5
Catalog No. 50-004-14315 Supplier Medchemexpress LLC Supplier No. 25GHYD0876
Add to Cart
Add to Cart
POPSO is a zwitterionic buffer that increases osmolality and demonstrates significant inhibition of anion uniport. It inhibits chloride uniport with an IC50 value of 24 mM. POPSO also enhances copper uptake and toxicity in alga and impairs the mitochondrial inner membrane. The working pH range for POPSO sesquisodium salt is 7.2-8.5.
  • Zwitterionic buffer
  • Increases osmolality
  • Marked inhibition of anion uniport
  • Inhibits chloride uniport with an IC50 value of 24 mM
  • Enhances copper uptake and toxicity in alga
  • Impairs mitochondrial inner membrane
  • Working pH range (for sesquisodium salt): 7.2-8.5